Learn More
1-(tert-butyl) 2-methyl 1H-indole-1,2-dicarboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
Formula Weight | 275.3 |
Percent Purity | 97% |
Chemical Name or Material | 1-(tert-Butyl) 2-methyl 1H-indole-1, 2-dicarboxylate |