missing translation for 'onlineSavingsMsg'
Learn More

1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7077422
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1190380-49-4
C8H13N3O
167.21
MFCD11505007
GZZNBQLBGSVOOZ-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine, 1-oxan-4-yl pyrazol-4-amine, 1-oxan-4-yl-1h-pyrazol-4-amine, 1h-pyrazol-4-amine, 1-tetrahydro-2h-pyran-4-yl, 1-tetrahydropyran-4-yl pyrazol-4-amine, 1-tetrahydro-pyran-4-yl-1h-pyrazol-4-ylamine, 1-2h-3,4,5,6-tetrahydropyran-4-yl pyrazole-4-ylamine
51063703
NC1=CN(N=C1)C1CCOCC1
This item is not returnable. View return policy
1190380-49-4
C8H13N3O
167.21
MFCD11505007
GZZNBQLBGSVOOZ-UHFFFAOYSA-N
1-tetrahydro-2h-pyran-4-yl-1h-pyrazol-4-amine, 1-oxan-4-yl pyrazol-4-amine, 1-oxan-4-yl-1h-pyrazol-4-amine, 1h-pyrazol-4-amine, 1-tetrahydro-2h-pyran-4-yl, 1-tetrahydropyran-4-yl pyrazol-4-amine, 1-tetrahydro-pyran-4-yl-1h-pyrazol-4-ylamine, 1-2h-3,4,5,6-tetrahydropyran-4-yl pyrazole-4-ylamine
51063703
NC1=CN(N=C1)C1CCOCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 105.0°C to 107.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 1-(oxan-4-yl)-1H-pyrazol-4-amine
Formula Weight 167.21
Percent Purity 97%
Chemical Name or Material 1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine
compliance-icons
Product Identifier
  • 1-Tetrahydropyran-4-yl-1H-pyrazol-4-ylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'