Läs mer
1-Tetratriacontanol, TRC
Produktkod. 17989734
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17989734 1 g
17928404 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17989734 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT3064801G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28484-70-0
Molekylformel C34 H70 O
Molekylvikt (g/mol) 494.92
Synonym Geddyl Alcohol, Sapiol, Tetratriacontyl Alcohol
IUPAC-namn tetratriacontan-1-ol
LEDER CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO
Specifikationer
Kemiskt namn eller material 1-Tetratriacontanol
Kvantitet 1 g
InChI formel InChI=1S/C34H70O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31-32-33-34-35/h35H,2-34H2,1H3
Formel vikt 494.5427