Learn More
[1-(Trimethylammonium)methyl] Methanethiosulfonate Bromide, TRC
Product Code. 30454908
Change view
Click to view available options
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30454908 1 g
30453723 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30454908 Supplier Toronto Research Chemicals Supplier No. TRCT7959201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 386229-81-8
Molekylformel C5 H14 N O2 S2 . Br
Molekylvikt (g/mol) 264.2
IUPAC-namn trimethyl(methylsulfonylsulfanylmethyl)azanium;bromide
LEDER [Br-].C[N+](C)(C)CSS(=O)(=O)C
Specifications
Kemiskt namn eller material (1-(Trimethylammonium)methyl) Methanethiosulfonate Bromide
Kvantitet 1 g
InChI formel InChI=1S/C5H14NO2S2.BrH/c1-6(2,3)5-9-10(4,7)8;/h5H2,1-4H3;1H/q+1;/p-1
Formel vikt 262.9649