Läs mer
10,11-Dihydro-11-[4-[2-(2-hydroxyethoxy)ethyl]-1-piperazinyl]dibenzo[b,f][1,4]thiazepin-11-ol, TRC
Produktkod. 30424754
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424754 100 mg
30415325 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30424754 Leverantör Toronto Research Chemicals Leverantörsnummer TRCQ510033100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 6-[4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]-5H-benzo[b][1,4]benzothiazepin-6-ol
CAS 1241800-24-7
Molekylformel C21 H27 N3 O3 S
Kvantitet 100 mg
Synonym Quetol
InChI formel InChI=1S/C21H27N3O3S/c25-14-16-27-15-13-23-9-11-24(12-10-23)21(26)17-5-1-3-7-19(17)28-20-8-4-2-6-18(20)22-21/h1-8,22,25-26H,9-16H2
LEDER OCCOCCN1CCN(CC1)C2(O)Nc3ccccc3Sc4ccccc24
IUPAC-namn 6-[4-[2-(2-hydroxyethoxy)ethyl]piperazin-1-yl]-5H-benzo[b][1,4]benzothiazepin-6-ol
Molekylvikt (g/mol) 401.522
Formel vikt 401.177