missing translation for 'onlineSavingsMsg'
Läs mer

10,11-Dihydrobenz[a]anthracene, TRC

Produktkod. 30374473
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374473 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30374473 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45034510MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 10,11-Dihydrobenz[a]anthracene
CAS 34501-50-3
Molekylformel C18H14
Kvantitet 10 mg
InChI formel InChI=1S/C18H14/c1-2-7-15-12-18-16(11-14(15)6-1)10-9-13-5-3-4-8-17(13)18/h1,3-6,8-12H,2,7H2
LEDER C1(C=C(C=CC2=C3C=CC=C2)C3=C4)=C4CCC=C1
IUPAC-namn 10,11-dihydrotetraphene
Molekylvikt (g/mol) 230.3
Formel vikt 230.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.