Learn More
10-[(2R)-3-(Dimethylamino)-2-methylpropyl]-2-methoxy-10H-phenothiazine 5-Oxide (Levomepromazine S-Oxide), Mikromol™
Description
10-[(2R)-3-(Dimethylamino)-2-methylpropyl]-2-methoxy-10H-phenothiazine 5-Oxide (Levomepromazine S-Oxide)
Specifications
Specifications
| Chemical Name or Material | Levomepromazine S-Oxide |
| CAS | 7606-29-3 |
| Analyte or Component Names | Levomepromazine S-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 O2 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Levomepromazine Maleate Imp. B (EP), Levomepromazine Imp. B (EP), 10-[(2R)-3-(Dimethylamino)-2-methylpropyl]-2-methoxy-10H-phenothiazine 5-Oxide, Levomepromazine S-Oxide, Levomepromazine Sulphoxide, (betaR)-2-Methoxy-N,N,beta-trimethyl-10H-phenothiazine-10-propanamine 5-oxide, (-)-10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine 5-oxide, Levomepromazine 5-sulfoxide, Levomepromazine sulfoxide, Levopromazine sulfoxide, Methotrimeprazine 5-sulfoxide, Methotrimeprazine sulfoxide, Levomepromazine Maleate Impurity B, Levomepromazine Impurity B |
| InChI Formula | InChI=1S/C19H24N2O2S/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)24(22)19-10-9-15(23-4)11-17(19)21/h5-11,14H,12-13H2,1-4H3/t14-,24?/m1/s1 |
| SMILES | COc1ccc2c(c1)N(C[C@H](C)CN(C)C)c3ccccc3S2=O |
| IUPAC Name | (2R)-3-(2-methoxy-5-oxophenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine |
| Show More |