missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ -10 to +360 Lo Tox™ Immersion Laboratory Thermometer

Lo Tox™ organic filling is specially chosen to achieve wide temperature span and provide the lowest practical levels of hazard.

601.00 SEK

Specifications

Length (Metric) 305 mm
Material Glass
Thermometer Fill Brown Organic Filling
Product Type Laboratory Immersion Thermometer
Max. Temperature (Metric) 360°C
View More Specs

Products 2
Product Code Brand Immersion Type Price Quantity & Availability  
Product Code Brand Immersion Type Price Quantity & Availability  
15432695
View Documents Promotion Details
Fisherbrand™
Total Immersion
601.00 SEK
Pack of 10
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15482695
View Documents Promotion Details
Fisherbrand™
Partial Immersion
601.00 SEK
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 10-05-2024
to see stock.
Add to basket
 
Description

Description

  • Quality and legibility make these an ideal substitute for mercury thermometers
  • Fitted with Brannan non roll end cap which has ring to allow the thermometer to be suspended
Specifications

Specifications

305 mm
Brown Organic Filling
360°C
-10°C to +360° C
Glass
Laboratory Immersion Thermometer
-10°C
±2°C at 90°C
Videos
SDS
Documents

Documents

Product Certifications