Läs mer
Tocris Bioscience™ 10-Cl-BBQ
Beskrivning
10-Cl-BBQ is a potent aryl hydrocarbon receptor (AhR) agonist, discovered as a screening hit (IC50 = 2.6 nM). Directly binds CD4+ T cells to induce AhR-dependent Tregs. Suppresses IL-17 production and prevents insulitis in NOD mice. In vivo half llife ∽ 2 hours, orally bioavailable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 10-Chloro-7H-benzimidazo[2,1-a]benz[de]isoquinolin-7-one |
| CAS | 23982-76-5 |
| Kvantitet | 5 mg |
| Molekylformel | C18H9ClN2O |
| Formel vikt | 304.73 |
| Renhet | 97% |