Läs mer
10-Deacetyl-7-xylosyl Paclitaxel (62%), TRC
Produktkod. 30362044
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362044 10 mg
30349969 50 mg
30351520 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362044 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD19896510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90332-63-1
Molekylformel C50 H57 N O17
Molekylvikt (g/mol) 943.98
Synonym Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-6,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-4-(β-D-xylopyranosyloxy)-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, (αR,βS)-, Benzenepropanoic acid, β-(benzoylamino)-α-hydroxy-, 12b-(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-6,11-dihydroxy-4a,8,13,13-tetramethyl-5-oxo-4-(β-D-xylopyranosyloxy)-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxet-9-yl ester, [2aR-[2aα,4β,4aβ,6β,9α(αR*,βS*),11α,12α,12aα,12bα]]-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete, benzenepropanoic acid deriv., 10-Deacetyl-7-xylosylpaclitaxel, 10-Deacetyl-7-xylosyltaxol, 10-Deacetylpaclitaxel 7-xyloside, 10-Deacetyltaxol 7-xyloside, 7-Xylosyl-10-deacetylpaclitaxel, 7-Xylosyl-10-deacetyltaxol, 7β-Xylosyl-10-deacetyltaxol
IUPAC-namn [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4-acetyloxy-15-[(2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoyl]oxy-1,12-dihydroxy-10,14,17,17-tetramethyl-11-oxo-9-[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
LEDER CC(=O)O[C@@]12CO[C@@H]1C[C@H](O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)[C@]4(C)[C@@H]2[C@H](OC(=O)c5ccccc5)[C@]6(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c7ccccc7)c8ccccc8)C(=C([C@@H](O)C4=O)C6(C)C)C
Specifikationer
Kemiskt namn eller material 10-Deacetyl-7-xylotaxol
Kvantitet 10 mg
InChI formel InChI=1S/C50H57NO17/c1-25-31(65-45(61)38(56)35(27-15-9-6-10-16-27)51-43(59)28-17-11-7-12-18-28)22-50(62)42(67-44(60)29-19-13-8-14-20-29)40-48(5,41(58)37(55)34(25)47(50,3)4)32(21-33-49(40,24-64-33)68-26(2)52)66-46-39(57)36(54)30(53)23-63-46/h6-20,30-33,35-40,42,46,53-57,62H,21-24H2,1-5H3,(H,51,59)/t30-,31+,32+,33-,35+,36+,37-,38-,39-,40+,42+,46+,48-,49+,50-/m1/s1
Formel vikt 943.3626