Läs mer
10-Hydroxy-10,11-dihydrocarbamazepine, Mikromol™
Produktkod. 30050743
Change view
Kvantitet:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050743 25 mg
17960406 100 mg
30459062 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30050743 Leverantör Mikromol Leverantörsnummer MM0076.100025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
10-Hydroxy-10,11-dihydrocarbamazepine
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 10,11-Dihydro-10-Hydroxycarbamazepine
CAS 29331-92-8
Analyt- eller komponentnamn 10,11-Dihydro-10-Hydroxycarbamazepine
Frakt skick Room Temperature
Molekylformel C15 H14 N2 O2
Certifikat och efterlevnad ISO 17025
Synonym 10-Hydroxy-10,11-dihydrocarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-hydroxycarbamazepine, 10-Hydroxy-10,11-dihydrocarbamezepine, BIA 2-005, GP 47779, Licarbazepine
InChI formel InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)
LEDER NC(=O)N1c2ccccc2CC(O)c3ccccc13
IUPAC-namn 5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Molekylvikt (g/mol) 254.28
Kvantitet 25 mg
Formel vikt 254.1055 g/mol