missing translation for 'onlineSavingsMsg'
Läs mer

10-Hydroxy-2-decanone, TRC

Product Code. 30406544
Change view
Click to view available options
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30406544 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406544 Supplier Toronto Research Chemicals Supplier No. TRCH943310100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 10-Hydroxy-2-decanone
CAS 35295-48-8
Molekylformel C10H20O2
Kvantitet 100 mg
InChI formel InChI=1S/C10H20O2/c1-10(12)8-6-4-2-3-5-7-9-11/h11H,2-9H2,1H3
LEDER CC(=O)CCCCCCCCO
IUPAC-namn 10-hydroxydecan-2-one
Molekylvikt (g/mol) 172.26
Formel vikt 172.1463
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.