missing translation for 'onlineSavingsMsg'
Läs mer

10-Hydroxyiminodibenzyl, Mikromol™

Produktkod. 30051403
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051403 100 mg
30052081 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051403 Supplier Mikromol Supplier No. MM0076.07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

10-Hydroxyiminodibenzyl
TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 10-Hydroxyiminodibenzyl
CAS 4014-77-1
Analyt- eller komponentnamn 10-Hydroxyiminodibenzyl
Frakt skick Room Temperature
Molekylformel C14 H13 N O
Certifikat och efterlevnad ISO 17025
Synonym 10,11-Dihydro-5H-dibenz[b,f]azepin-10-ol, 10-Hydroxyiminodibenzyl
InChI formel InChI=1S/C14H13NO/c16-14-9-10-5-1-3-7-12(10)15-13-8-4-2-6-11(13)14/h1-8,14-16H,9H2
LEDER OC1Cc2ccccc2Nc3ccccc13
IUPAC-namn 6,11-dihydro-5H-benzo[b][1]benzazepin-5-ol
Molekylvikt (g/mol) 211.26
Kvantitet 100 mg
Formel vikt 211.0997 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.