Läs mer
Tocris Bioscience™ VU 0650991
GREENER_CHOICE
Produktkod. 18749186
Kvantitet:
10 mg
50 mg
Produktkod. 18749186
Brand: Tocris Bioscience™ 6355/10
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Non-competitive GLP-1 antagonist; orally bioavailable and brain penetrant
VU 0650991 is a noncompetitive glucagon-like peptide-1 (GLP-1) receptor antagonist (IC50 = 0.65 - 0.69 μM). Displays >40-fold selectivity for GLP-1 receptor over glucagon receptors. Blocks activity of Exendin-4 (Cat. No. 1933) in potentiating insulin secretion from primary mouse pancreatic islets in vitro. Decreases blood insulin and increases blood glucose levels in vivo. Brain penetrant and orally bioavailable.
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 10 mg
Molekylformel C16H11ClF6N4O2
Formel vikt 440.73
Produkttyp Antagonist
Renhet 98%