Läs mer
10-trans,12-cis-Linoleic Acid, TRC
Produktkod. 30476702
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30476702 50 mg
1 options
Produktkod. 30476702 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL46252050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 10-trans,12-cis-Linoleic Acid
CAS 2420-56-6
Molekylformel C18 H32 O2
Kvantitet 50 mg
Synonym 10,12-Octadecadienoic acid, (Z,E)-, (10E,12Z)-10,12-Octadecadienoic acid, 10-trans,12-cis-Linoleic acid, 10-trans-12-cis-Octadecadienoic acid, 10E,12Z-Octadecadienoic acid, 10t12c Conjugated linoleic acid, C18:2 t-10,c-12, trans-10,cis-12 Conjugated linoleic acid, trans-10-cis-12-Octadecadienoic acid, 10,12-Octadecadienoic acid, (10E,12Z)-, (10E,12Z)-10,12-Octadecadienoic Acid
InChI formel InChI=1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-9H,2-5,10-17H2,1H3,(H,19,20)/b7-6-,9-8+
LEDER CCCCC\C=C/C=C/CCCCCCCCC(=O)O
IUPAC-namn (10E,12Z)-octadeca-10,12-dienoic acid
Molekylvikt (g/mol) 280.45
Formel vikt 280.2402