Läs mer
10-Undecenoic Acid Ethyl Ester, TRC
Produktkod. 30450977
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30450977 25 g
1 options
Produktkod. 30450977 Leverantör Toronto Research Chemicals Leverantörsnummer TRCU78882525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 10-Undecenoic Acid Ethyl Ester
CAS 692-86-4
Molekylformel C13H24O2
Kvantitet 25 g
Synonym Ethyl 10-Undecenoate, Ethyl Undecenoate
InChI formel InChI=1S/C13H24O2/c1-3-5-6-7-8-9-10-11-12-13(14)15-4-2/h3H,1,4-12H2,2H3
LEDER CCOC(=O)CCCCCCCCC=C
IUPAC-namn ethyl undec-10-enoate
Molekylvikt (g/mol) 212.33
Formel vikt 212.1776