Beskrivning
S100A5 Monoclonal antibody specifically detects S100A5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifikationer
Specifikationer
| Antigen | S100A5 |
| Användningsområden | Immunohistochemistry (Paraffin) |
| Klassificering | Monoclonal |
| Klona | S100A5/7472 |
| Koncentration | 0.2 mg/mL |
| Konjugera | Unconjugated |
| Utspädning | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Formulering | 10 mM PBS with 0.05% BSA |
| Gene Alias | Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 |
| Värd art | Mouse |
| Visa mer |