Learn More
1LT Buffer Solution, pH 9.00 [Borate] BAKER ANALYZED
Brand:  JT Baker 7145.1000
1LT Buffer Solution, pH 9.00 [Borate] BAKER ANALYZED
Additional Details : Weight : 1.00000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Product Code. 10341801
  • 665.00 SEK / 1 L
Estimated Shipment: 15-05-2024
to see stock.
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.