Learn More
Methylamine, 33 wt.% solution in absolute ethanol, AcroSeal(R), Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 396731000
Additional Details : CAS Number : 74-89-5 Weight : 0.07560kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methylamine | |
0.7560 g/mL | |
AcroSeal™ Glass bottle | |
CH3NH2 | |
0.756 | |
Solubility aq. soln.: soluble. | |
31.06 |
74-89-5 | |
-23°C | |
CH5N | |
100 mL | |
15, 6088 | |
31.06 |
Safety and Handling
- Methylamine, 33 wt% solution in absolute ethanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
For Manufacturing and Laboratory Use. Not for beverage use