Learn More
Keystone Cleanroom Products 1073B Tyvek™ Autoclaveable Wrapper Small
BIOPHARMAGREENER_CHOICE
Provides an excellent bacterial barrier for superior performance post steam sterilization. Cut using an automated CAD system in ISO 8 and made from DuPont 1073B Medical Grade Tyvek.
Brand:  Keystone Cleanroom Products MGTVKSHEET0836
 View more versions of this product
Product Code. 17758447
  • 15945.00 SEK / Pack of 500
Load Up On Autoclave Essentials
Use promo code 24153 at checkout
View Available Promotions
Only null left
Estimated Shipment: 22-08-2025
Description
  • Designed for Steam Sterilization
  • Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
  • Hydrophobic
  • Excellent Microbial Barrier
  • Resists Tearing and Punctures
  • Lint-Free - Category (Helmke Drum)
  • Manufactured and Packaged in a ISO 6 Cleanroom
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclavable
None
Single-use
Pharmaceutical Equipment
Non-sterile
Steam
<125°C
36 in.
8 in.
Sterilization Wrap
COC
White
Steam Sterilization
Tyvek
Yes
<250°F
36 x 8 in.
91.4 cm
20.3 cm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Keystone Cleanroom Products 1073B Tyvek™ Autoclaveable Wrapper Small > Size (WxL): 8 x 36 in.