missing translation for 'onlineSavingsMsg'
Läs mer

3-Hydroxy Donepezil (Donepezil Impurity), TRC

Produktkod. 17993965
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17993965 10 mg
30137451 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17993965 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94174510MG

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2097683-67-3
Kemiskt namn eller material 3-Hydroxy Donepezil
Formel vikt 395.2097
InChI formel InChI=1S/C24H29NO4/c1-28-21-13-18-19(14-22(21)29-2)24(27)20(23(18)26)12-16-8-10-25(11-9-16)15-17-6-4-3-5-7-17/h3-7,13-14,16,20,23,26H,8-12,15H2,1-2H3
IUPAC-namn 2-[(1-benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxy-2,3-dihydroinden-1-one
Molekylformel C24 H29 N O4
Molekylvikt (g/mol) 395.49
Procent renhet >95%
Anteckningar om renhetsgrad HPLC
LEDER COc1cc2C(O)C(CC3CCN(Cc4ccccc4)CC3)C(=O)c2cc1OC
Synonym 1H-Inden-1-one, 2,3-dihydro-3-hydroxy-5,6-dimethoxy-2-[[1-(phenylmethyl)-4-piperidinyl]methyl]-, 3-Hydroxydonepezil, 2-[(1-Benzylpiperidin-4-yl)methyl]-3-hydroxy-5,6-dimethoxyindan-1-one
Show More Show Less

Specifications

Kvantitet 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.