Läs mer
Novus Biologicals™ 10nm Streptavidin Gold Conjugate (10OD)
Produktkod. 18630281 Handla allt Bio Techne Produkter
Kvantitet:
0.2 mL
1 mL
Produktkod. 18630281
Brand: Novus Biologicals™ 2521000
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Streptavidin gold nanoparticle conjugates can be used in conjunction with biotinylated reagents such as biotinylated antibodies or proteins, in applications such as lateral flow immunoassays or electron microscopy.
InnovaCoat GOLD nanoparticles have a proprietary surface coating which covalently binds antibodies, proteins, peptides or any other biomolecule with free amine groups to form highly stable conjugates. The surface coating can withstand the most extreme conditions (e.g. 2.5M NaOH at 70°C for 2 hours), making InnovaCoat™ GOLD an ideal reagent for a wide range of applications.
TRUSTED_SUSTAINABILITY
Specifikationer
Kvantitet 1 mL
Innehåll och lagring 4°C Do not freeze.