Läs mer
11,11′-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride, Mikromol™
Beskrivning
11,11′-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | 11,11'-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride |
| CAS | 1923194-81-3 |
| Analyt- eller komponentnamn | 11,11'-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C40 H44 N6 O2 S2 . 4 Cl H |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | 1,2-Bis[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethane Tetrahydrochloride, 11,11-2-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride, Quetiapine Fumarate Imp. E (EP) as Tetrahydrochloride, 11,11′-[Ethylenebis(oxyethylenepiperazine-4,1-diyl)]bis(dibenzo[b,f][1,4]thiazepine) Tetrahydrochloride, Quetiapine Fumarate Impurity E as Tetrahyhydrochloride, Quetiapine Impurity E as Tetrahyhydrochloride |
| InChI formel | InChI=1S/C40H44N6O2S2.4ClH/c1-5-13-35-31(9-1)39(41-33-11-3-7-15-37(33)49-35)45-21-17-43(18-22-45)25-27-47-29-30-48-28-26-44-19-23-46(24-20-44)40-32-10-2-6-14-36(32)50-38-16-8-4-12-34(38)42-40;;;;/h1-16H,17-30H2;4*1H |
| LEDER | Cl.Cl.Cl.Cl.C(COCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24)OCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68 |
| IUPAC-namn | 6-[4-[2-[2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethoxy]ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepine;tetrahydrochloride |
| Visa mer |