Läs mer
11-Beta,16-Alpha,17-Alpha,21-Tetrahydroxypregna-1,4-diene-3,20-dione, TRC
Produktkod. 30009379
Change view
Kvantitet:
50 mg
500 mg
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009379 10 g
30007540 500 mg
17973155 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009379 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29607510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13951-70-7
Molekylformel C21 H28 O6
Molekylvikt (g/mol) 376.44
Synonym 11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione, Budesonide Imp. A (EP), 16alpha-Hydroxyprednisolone;
IUPAC-namn (8S,9S,10R,11S,13S,14S,16R,17S)-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 16alpha-Hydroxyprednisolone (11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 g
InChI formel InChI=1S/C21H28O6/c1-19-6-5-12(23)7-11(19)3-4-13-14-8-16(25)21(27,17(26)10-22)20(14,2)9-15(24)18(13)19/h5-7,13-16,18,22,24-25,27H,3-4,8-10H2,1-2H3/t13-,14-,15-,16+,18+,19-,20-,21-/m0/s1
Formel vikt 376.1886
Procent renhet >95%