Läs mer
11-Dehydrocorticosterone, TRC
Produktkod. 30137700
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137700 50 mg
30006505 5 mg
30012564 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23011550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72-23-1
Molekylformel C21 H28 O4
Molekylvikt (g/mol) 344.44
Synonym Pregn-4-ene-3,11,20-trione, 21-hydroxy-, 21-Hydroxypregn-4-ene-3,11,20-trione, 11-Dehydrocorticosteron, 11-Dehydrocorticosterone, 11-Oxo-11-deoxycorticosterone, 11-Oxocorticosterone, 17-(1-keto-2-Hydroxyethyl)-δ4-androsten-3,11-dione, 4-Pregnen-21-ol-3,11,20-trione, Corticosterone, 11-dehydro-, Dehydrocorticosterone, Kendall's compound A, NSC-9702, δ4-Pregnen-21-ol-3,11,20-trione, δ4-Pregnene-21-ol-3,11,20-trione
IUPAC-namn (8S,9S,10R,13S,14S,17S)-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,12,14,15,16,17-decahydro-1H-cyclopenta[a]phenanthrene-3,11-dione
LEDER C[C@]12CC(=O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO
Specifikationer
Kemiskt namn eller material 11-Dehydrocorticosterone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C21H28O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h9,14-16,19,22H,3-8,10-11H2,1-2H3/t14-,15-,16+,19+,20-,21-/m0/s1
Formel vikt 344.1988
Procent renhet >95%