Läs mer
11-Deoxy Cortisol, TRC
Produktkod. 18082240
Change view
Kvantitet:
5 mg
10 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18082240 5 mg
18090340 10 mg
18033950 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18082240 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2326005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 152-58-9
Molekylformel C21 H30 O4
Molekylvikt (g/mol) 346.46
Synonym 17,21-Dihydroxypregn-4-ene-3,20-dione, Hydrocortisone Imp. F (EP), Reichstein's Substance S, 11-Deoxycortisol, Cortexolone, Pregn-4-ene-3,20-dione, 17,21-dihydroxy-, Compound S (6CI), 11-Deoxy-17-hydrocorticosterone, 11-Deoxy-17-hydroxycorticosterone, 11-Deoxycortisone, 11-Deoxyhydrocortisone, 11-Desoxy-17α-hydroxycorticosterone, 11-Desoxycortisol, 11-Desoxyhydrocortisone, 17,21-Dihydroxypregn-4-en-3,20-dione, 17,21-Dihydroxyprogesterone, 17-Hydroxy-11-deoxycorticosterone, 17α,21-Dihydroxy-4-pregnen-3,20-dione, 17α,21-Dihydroxypregn-4-ene-3,20-dione, 17α,21-Dihydroxyprogesterone, 17α-Hydroxycortexone, 4-Pregnen-17α,21-diol-3,20-dione, 4-Pregnene-17α,21-diol-3,20-dione, Cortifen, Cortisol, 11-deoxy-, Cortodoxone, NSC 18317, Reichstein S, Reichstein substance S, Reichstein's compound S, KF 3050, Substance S, δ4-Pregnene-17α,21-diol-3,20-dione
IUPAC-namn (8R,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12CCC(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 11-Deoxycortisol
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C21H30O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h11,15-17,22,25H,3-10,12H2,1-2H3/t15-,16+,17+,19+,20+,21+/m1/s1
Formel vikt 346.2144
Procent renhet >95%