Learn More
11-Deoxy Fusidic Acid, TRC
Product Code. 30366037
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Product Code. Quantity Unit Size
30366037 2.5 mg 2.5mg
1 options
Product Code. 30366037 Supplier Toronto Research Chemicals Supplier No. TRCD2363002.5MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 11-Deoxyfusidic Acid
CAS 1013937-16-0
Molecular Formula C31 H48 O5
Quantity 2.5 mg
Synonym 29-Nordammara-17(20),24-dien-21-oic acid, 16-(acetyloxy)-3-hydroxy-, (3α,4α,8α,9β,13α,14β,16β,17Z)-, (3α,4α,8α,9β,13α,14β,16β,17Z)-16-(Acetyloxy)-3-hydroxy-29-nordammara-17(20),24-dien-21-oic acid, ent-(17Z)-16α-(Acetyloxy)-3β-hydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dien-21-oic acid, 11-Deoxyfusidic acid
InChI Formula InChI=1S/C31H48O5/c1-18(2)9-8-10-21(28(34)35)27-23-11-12-26-29(5)15-14-24(33)19(3)22(29)13-16-30(26,6)31(23,7)17-25(27)36-20(4)32/h9,19,22-26,33H,8,10-17H2,1-7H3,(H,34,35)/b27-21-/t19-,22-,23-,24+,25-,26-,29-,30-,31-/m0/s1
SMILES C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2CC[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](C[C@]34C)OC(=O)C
IUPAC Name (2Z)-2-[(3R,4S,5S,8S,9S,10S,13R,14S,16S)-16-acetyloxy-3-hydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]-6-methylhept-5-enoic acid
Molecular Weight (g/mol) 500.7098
Formula Weight 500.3502