Läs mer
11-Deoxy Prednisolone, TRC
Produktkod. 18010250
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18010250 50 mg
18080540 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18010250 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD24985050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1807-14-3
Molekylformel C21 H28 O4
Molekylvikt (g/mol) 344.44
Synonym 17,21-Dihydroxy-pregna-1,4-diene-3,20-dione, 11-Deoxyprednisolone, 17,21-Dihydroxypregna-1,4-diene-3,20-dione, 17α,21-Dihydroxypregn-1,4-dien-3,20-dione, 17α,21-Dihydroxypregna-1,4-diene-3,20-dione, δ1,4-Reichstein S, δ'-Dehydrocortexolone
IUPAC-namn (8R,9S,10R,13S,14S,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 11-Deoxy Prednisolone
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C21H28O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h5,8,11,15-17,22,25H,3-4,6-7,9-10,12H2,1-2H3/t15-,16+,17+,19+,20+,21+/m1/s1
Formel vikt 344.1988
Procent renhet >95%