missing translation for 'onlineSavingsMsg'
Learn More

11-Deoxycorticosterone acetate, 97%, Thermo Scientific Chemicals

Catalog Number p-7077839
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
56-47-3
C23H32O4
372.51
MFCD00003660
VPGRYOFKCNULNK-ZIUYWEQENA-N
deoxycorticosterone acetate, desoxycorticosterone acetate, doca, desoxycortone acetate, percotol, cortexone acetate, decosteron, decosterone, sincortex, syncortyl
5952
CHEBI:34671
[H][C@@]12CC[C@H](C(=O)COC(C)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])CCC2=CC(=O)CC[C@]12C
This item is not returnable. View return policy
56-47-3
C23H32O4
372.51
MFCD00003660
VPGRYOFKCNULNK-ZIUYWEQENA-N
deoxycorticosterone acetate, desoxycorticosterone acetate, doca, desoxycortone acetate, percotol, cortexone acetate, decosteron, decosterone, sincortex, syncortyl
5952
CHEBI:34671
[H][C@@]12CC[C@H](C(=O)COC(C)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])CCC2=CC(=O)CC[C@]12C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 154.0°C to 160.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name [2-[(8S,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Formula Weight 372.5
Percent Purity 97%
Chemical Name or Material 11-Deoxycorticosterone acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'