Läs mer
11-Keto Budesonide (Mixture of Diastereomers), TRC
Produktkod. 17981215
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17981215 100 mg
17962925 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17981215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK175250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 216453-74-6
Molekylformel C25 H32 O6
Molekylvikt (g/mol) 428.52
Synonym 16α,17-[(1RS)-Butylidene-bis(oxy)]-21-hydroxypregna-1,4-diene-3,11,20-trione, Budesonide Imp. L (EP), Budesonide Related Compound L, Budesonide Impurity L
IUPAC-namn (1S,2S,4R,8S,9S,12S,13R)-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-diene-11,16-dione
LEDER CCCC1O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4C(=O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 11-Keto Budesonide
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C25H32O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-10,16-17,20-22,26H,4-7,11-13H2,1-3H3/t16-,17-,20+,21?,22+,23-,24-,25+/m0/s1
Formel vikt 428.2199
Procent renhet >95%