Läs mer
11-Keto Fusidic Acid, TRC
Produktkod. 17942055
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17942055 10 mg
17931305 50 mg
17931535 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17942055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK18899010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 16711-91-4
Molekylformel C31H46O6
Molekylvikt (g/mol) 514.69
Synonym (3α,4α,8α,9β,13α,14β,16β,17Z)-16-(Acetyloxy)-3-hydroxy-11-oxo-29-nordammara-17(20),24-dien-21-oic Acid, 11-Oxofusidic Acid, Fusidic Acid EP Impurity H
LEDER O=C1C[C@](/C([C@@H](OC(C)=O)C2)=C(C(O)=O)\CC/C=C(C)/C)([H])[C@@]2(C)[C@@]3(C)CC[C@@]4([H])[C@H](C)[C@H](O)CC[C@]4(C)[C@@]31[H]
Specifikationer
Kemiskt namn eller material 11-Keto Fusidic Acid