Läs mer
11-Mercapto-1-undecanol, TRC
Produktkod. 30483637
Change view
Kvantitet:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30483637 1 g
30469189 5 g
30473924 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483637 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2575851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73768-94-2
Molekylformel C11H24OS
Molekylvikt (g/mol) 204.37
Synonym 11-Mercaptoundecyl Alcohol, 11-Sulfanylundecan-1-ol, ω-Mercaptoundecanol, 11-Hydroxyl-1-undecylthiol, 11-Hydroxy-1-undecanethiol, 1-Mercaptoundecan-11-ol
IUPAC-namn 11-sulfanylundecan-1-ol
LEDER OCCCCCCCCCCCS
Specifikationer
Kemiskt namn eller material 11-Mercapto-1-undecanol
Kvantitet 1 g
InChI formel InChI=1S/C11H24OS/c12-10-8-6-4-2-1-3-5-7-9-11-13/h12-13H,1-11H2
Formel vikt 204.1548