Läs mer
11-Oxo Androsterone, TRC
Produktkod. 30135707
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135707 5 mg
30005541 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135707 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8471105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1231-82-9
Molekylformel C19 H28 O3
Molekylvikt (g/mol) 304.42
Synonym Androstane-11,17-dione, 3-hydroxy-, (3α,5α)-, 5α-Androstane-11,17-dione, 3α-hydroxy- (8CI), (3α,5α)-3-Hydroxyandrostane-11,17-dione, 11-Oxoandrosterone, 3α-Hydroxy-5α-androstan-11,17-dione, 3α-Hydroxy-5α-androstane-11,17-dione, 5α-Androstan-3α-ol-11,17-dione, 5α-Androstane-3α-ol-11,17-dione, Ba 2683, NSC 56412
IUPAC-namn (3R,5S,8S,9S,10S,13S,14S)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthrene-11,17-dione
LEDER C[C@]12CC[C@@H](O)C[C@@H]1CC[C@H]3[C@@H]4CCC(=O)[C@@]4(C)CC(=O)[C@H]23
Specifikationer
Kemiskt namn eller material 11-Oxo Androsterone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C19H28O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h11-14,17,20H,3-10H2,1-2H3/t11-,12+,13-,14-,17+,18-,19-/m0/s1
Formel vikt 304.2038
Procent renhet >95%