Läs mer
11-Oxo-betamethasone Dipropionate, TRC
Produktkod. 30134792
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134792 500 mg
30006482 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134792 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO869860500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 64967-90-4
Molekylformel C28H35FO7
Molekylvikt (g/mol) 502.57
Synonym Pregna-1,4-diene-3,11,20-trione, 9-fluoro-16-methyl-17,21-bis(1-oxopropoxy)-, (16β)- (9CI)
IUPAC-namn [2-[(8S,9R,10S,13S,14S,16S,17R)-9-fluoro-10,13,16-trimethyl-3,11-dioxo-17-propanoyloxy-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
LEDER CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C
Specifikationer
Kemiskt namn eller material 11-Oxo-betamethasone Dipropionate
Kvantitet 500 mg
InChI formel InChI=1S/C28H35FO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-20H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,25-,26-,27-,28-/m0/s1
Formel vikt 502.2367