Läs mer
(11Beta,16Alpha)-11-Hydroxy-16-(1-oxobutoxy)-androsta-1,4-diene-3,17-dione, TRC
Produktkod. 17962605
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17962605 10 mg
17920445 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17962605 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH82571510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85234-64-6
Molekylformel C23 H30 O5
Molekylvikt (g/mol) 386.48
Synonym Androsta-1,4-diene-3,17-dione, 11-hydroxy-16-(1-oxobutoxy)-, (11β,16α)-, (11β,16α)-11-Hydroxy-16-(1-oxobutoxy)androsta-1,4-diene-3,17-dione, 11β,16α-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate
IUPAC-namn [(8S,9S,10R,11S,13S,14S,16R)-11-hydroxy-10,13-dimethyl-3,17-dioxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-16-yl] butanoate
LEDER CCCC(=O)O[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]2(C)C1=O
Specifikationer
Kemiskt namn eller material 11beta,16alpha-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C23H30O5/c1-4-5-19(26)28-18-11-16-15-7-6-13-10-14(24)8-9-22(13,2)20(15)17(25)12-23(16,3)21(18)27/h8-10,15-18,20,25H,4-7,11-12H2,1-3H3/t15-,16-,17-,18+,20+,22-,23-/m0/s1
Formel vikt 386.2093
Procent renhet >95%