Läs mer
11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione (16alpha-Hydroxyprednisolone), Mikromol™
Produktkod. 30050653
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050653 25 mg
16891520 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050653 Leverantör Mikromol Leverantörsnummer MM0204.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione (16alpha-Hydroxyprednisolone)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 16alpha-Hydroxyprednisolone (11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione)
CAS 13951-70-7
Analyt- eller komponentnamn 16alpha-Hydroxyprednisolone (11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione)
Frakt skick Ice pack (-20°C)
Molekylformel C21 H28 O6
Certifikat och efterlevnad ISO 17025
Synonym 11beta,16alpha,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione, Budesonide Imp. A (EP), 16alpha-Hydroxyprednisolone, Budesonide Impurity A
InChI formel InChI=1S/C21H28O6/c1-19-6-5-12(23)7-11(19)3-4-13-14-8-16(25)21(27,17(26)10-22)20(14,2)9-15(24)18(13)19/h5-7,13-16,18,22,24-25,27H,3-4,8-10H2,1-2H3/t13-,14-,15-,16+,18+,19-,20-,21-/m0/s1
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1C[C@@H](O)[C@]2(O)C(=O)CO
IUPAC-namn (8S,9S,10R,11S,13S,14S,16R,17S)-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 376.44
Kvantitet 25 mg
Formel vikt 376.1886 g/mol