missing translation for 'onlineSavingsMsg'
Läs mer

11beta,16alpha-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate, Mikromol™

Produktkod. 17960646
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960646 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17960646 Leverantör Mikromol Leverantörsnummer MM0204.100025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

11beta,16alpha-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate
TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 11beta,16alpha-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate
CAS 85234-64-6
Analyt- eller komponentnamn 11beta,16alpha-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate
Frakt skick Room Temperature
Molekylformel C23 H30 O5
Certifikat och efterlevnad ISO 9001
Synonym Androsta-1,4-diene-3,17-dione, 11-hydroxy-16-(1-oxobutoxy)-, (11β,16α)-, (11β,16α)-11-Hydroxy-16-(1-oxobutoxy)androsta-1,4-diene-3,17-dione, 11β,16α-Dihydroxyandrosta-1,4-diene-3,17-dione 16-Butyrate
InChI formel InChI=1S/C23H30O5/c1-4-5-19(26)28-18-11-16-15-7-6-13-10-14(24)8-9-22(13,2)20(15)17(25)12-23(16,3)21(18)27/h8-10,15-18,20,25H,4-7,11-12H2,1-3H3/t15-,16-,17-,18+,20+,22-,23-/m0/s1
LEDER CCCC(=O)O[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]2(C)C1=O
IUPAC-namn [(8S,9S,10R,11S,13S,14S,16R)-11-hydroxy-10,13-dimethyl-3,17-dioxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-16-yl] butanoate
Molekylvikt (g/mol) 386.48
Kvantitet 25 mg
Formel vikt 386.2093 g/mol
Visa mer Visa mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.