Läs mer
(11beta)-21-Chloro-9-fluoro-11-hydroxy-16-methylpregna-1,4,16-triene-3,20-dione, TRC
Produktkod. 18050190
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18050190 50 mg
17949929 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18050190 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36632550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1356190-17-4
Molekylformel C22 H26 Cl F O3
Molekylvikt (g/mol) 392.89
Synonym (11beta)-21-Chloro-9-fluoro-11-hydroxy-16-methylpregna-1,4,16-triene-3,20-dione, 21-Chloro-9-fluoro-11beta-hydroxy-16-methylpregna-1,4,16-triene-3,20-dione, GR 156421X, Ph Eur Clobetasol Propionate Impurity B, Clobetasol, CCI18747, CCI6990, CCI1822, 21-desoxybetamethasone-17-propionate, 16-ene, Ethylenic acid, USP Clobetasol propionate Related Compound A
IUPAC-namn (8S,9R,10S,11S,13S,14S)-17-(2-chloroacetyl)-9-fluoro-11-hydroxy-10,13,16-trimethyl-7,8,11,12,14,15-hexahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER CC1=C(C(=O)CCl)[C@@]2(C)C[C@H](O)[C@@]3(F)[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]2C1
Specifikationer
Kemiskt namn eller material (11beta)-21-Chloro-9-fluoro-11-hydroxy-16-methylpregna-1,4,16-triene-3,20-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C22H26ClFO3/c1-12-8-16-15-5-4-13-9-14(25)6-7-21(13,3)22(15,24)18(27)10-20(16,2)19(12)17(26)11-23/h6-7,9,15-16,18,27H,4-5,8,10-11H2,1-3H3/t15-,16-,18-,20-,21-,22-/m0/s1
Formel vikt 392.1555
Procent renhet >95%