Läs mer
(11E)-Tetradecen-1-yl Acetate, TRC
Produktkod. 30415098
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30415098 100 mg
1 options
Produktkod. 30415098 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT291750100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (E)-11-Tetradecen-1-yl acetate
CAS 33189-72-9
Molekylformel C16 H30 O2
Kvantitet 100 mg
Synonym 11-Tetradecen-1-ol','acetate','(11E)- (9CI), 11-Tetradecen-1-ol','acetate','(E)- (8CI), (E)-11-Tetradecen-1-ol acetate, (E)-11-Tetradecen-1-yl acetate, (E)-11-Tetradecenyl acetate, 11E-Tetradecenyl acetate, trans-11-Tetradecen-1-ol acetate, trans-11-Tetradecen-1-yl acetate, trans-11-Tetradecenol acetate, trans-11-Tetradecenyl acetate
InChI formel InChI=1S/C16H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-16(2)17/h4-5H,3,6-15H2,1-2H3/b5-4+
LEDER CC\C=C\CCCCCCCCCCOC(=O)C
IUPAC-namn [(E)-tetradec-11-enyl] acetate
Molekylvikt (g/mol) 254.41
Formel vikt 254.2246