Läs mer
12-Aminolauric Acid, TRC
Produktkod. 30321638
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30321638 5 g
1 options
Produktkod. 30321638 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6122305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 12-Aminododecanoic acid
CAS 693-57-2
Molekylformel C12 H25 N O2
Kvantitet 5 g
Synonym 12-Aminododecanoic acid, 12-Amindodecanoic acid, 12-Aminolauric acid, ADA, λ-Aminolauric acid, ω-Aminododecanoic acid, ω-Aminolauric acid, Dodecanoic acid, 12-amino-, 12-Aminododecanoic Acid
InChI formel InChI=1S/C12H25NO2/c13-11-9-7-5-3-1-2-4-6-8-10-12(14)15/h1-11,13H2,(H,14,15)
LEDER NCCCCCCCCCCCC(=O)O
IUPAC-namn 12-aminododecanoic acid
Molekylvikt (g/mol) 215.33
Formel vikt 215.1885