missing translation for 'onlineSavingsMsg'
Learn More

12-Azidododecylphosphonic acid, 95%

Product Code. 15416647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416647 1 g 1g
15426647 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416647 Supplier Thermo Scientific Alfa Aesar Supplier No. H33784.03
 Restricted Item
Beräknad leverans: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 721457-32-5 | C12H26N3O3P | 291.332 g/mol

This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This coupling agent specifically reacts with alkyne functions to form the corresponding 1,2,3-triazole via a copper-free 1,3-dipolar cycloaddition.

Notes
Store in refrigerator. Protect from heat. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 721457-32-5
Molecular Formula C12H26N3O3P
Molecular Weight (g/mol) 291.332
MDL Number MFCD17012690
InChI Key WHRLXOVKIALVOF-UHFFFAOYSA-N
PubChem CID 97638952
IUPAC Name 12-azidododecylphosphonic acid
SMILES C(CCCCCCP(=O)(O)O)CCCCCN=[N+]=[N-]

Specifications

Melting Point 83°C to 85°C
Linear Formula N3(CH2)12P(O)(OH)2
Quantity 1 g
Formula Weight 291.33
Percent Purity 95%
Chemical Name or Material 12-Azidododecylphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.