Läs mer
12-Chlorododecanoic Acid, TRC
Produktkod. 17961675
Change view
Kvantitet:
500 mg
2.5 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17961675 5 g
17933685 500 mg
17900355 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17961675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3631655G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22075-86-1
Molekylformel C12H23ClO2
Molekylvikt (g/mol) 234.76
Synonym NSC 667272;
IUPAC-namn 12-chlorododecanoic acid
LEDER OC(=O)CCCCCCCCCCCCl
Specifikationer
Kemiskt namn eller material 12-Chlorododecanoic Acid
Kvantitet 5 g
InChI formel InChI=1S/C12H23ClO2/c13-11-9-7-5-3-1-2-4-6-8-10-12(14)15/h1-11H2,(H,14,15)
Formel vikt 234.1387