Läs mer
12-Hydroxylauric Acid, TRC
Produktkod. 30406700
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30406700 10 g
1 options
Produktkod. 30406700 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94377510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 12-Hydroxylauric Acid
CAS 505-95-3
Molekylformel C12 H24 O3
Kvantitet 10 g
Synonym 12-Hydroxydodecanoic Acid, NSC 159293, NSC 664211, ω-Hydroxydodecanoic Acid, ω-Hydroxylauric Acid
InChI formel InChI=1S/C12H24O3/c13-11-9-7-5-3-1-2-4-6-8-10-12(14)15/h13H,1-11H2,(H,14,15)
LEDER OCCCCCCCCCCCC(=O)O
IUPAC-namn 12-hydroxydodecanoic acid
Molekylvikt (g/mol) 216.32
Formel vikt 216.1725