Läs mer
12-Ketoursodeoxycholic Acid, TRC
Produktkod. 30134495
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134495 5 mg
30133683 1 mg
30004591 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK3154005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81873-91-8
Molekylformel C24 H38 O5
Molekylvikt (g/mol) 406.56
Synonym (3α,5β,7β)-3,7-Dihydroxy-12-oxo-cholan-24-oic Acid, SF 482
IUPAC-namn (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-12-oxo-1,2,3,4,5,6,7,8,9,11,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-17-yl]pentanoic acid
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC(=O)[C@]12C
Specifikationer
Kemiskt namn eller material 12-Ketoursodeoxycholic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C24H38O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-19,22,25-26H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19+,22+,23+,24-/m1/s1
Formel vikt 406.2719
Procent renhet >95%