Läs mer
12-Methyltridecanal, TRC
Produktkod. 30481960
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30481960 1 g
30466504 10 g
30480022 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30481960 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3311601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75853-49-5
Molekylformel C14 H28 O
Molekylvikt (g/mol) 212.37
IUPAC-namn 12-methyltridecanal
LEDER CC(C)CCCCCCCCCCC=O
Specifikationer
Kemiskt namn eller material 12-Methyltridecanal
Kvantitet 1 g
InChI formel InChI=1S/C14H28O/c1-14(2)12-10-8-6-4-3-5-7-9-11-13-15/h13-14H,3-12H2,1-2H3
Formel vikt 212.214