Läs mer
12-Pentacosanone, TRC
Produktkod. 30424082
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424082 25 mg
30423093 250 mg
30427500 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424082 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27026025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22026-18-2
Molekylformel C25H50O
Molekylvikt (g/mol) 366.66
Synonym Pentacosan-12-one
IUPAC-namn pentacosan-12-one
LEDER CCCCCCCCCCCCCC(=O)CCCCCCCCCCC
Specifikationer
Kemiskt namn eller material 12-Pentacosanone
Kvantitet 25 mg
InChI formel InChI=1S/C25H50O/c1-3-5-7-9-11-13-14-16-18-20-22-24-25(26)23-21-19-17-15-12-10-8-6-4-2/h3-24H2,1-2H3
Formel vikt 366.3862