missing translation for 'onlineSavingsMsg'
Läs mer

12Beta-deoxikolsyra metylester 3Alfa-bensoat, TRC

Product Code. 30366906
Change view
Click to view available options
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30366906 100 mg
30364602 50 mg
30365603 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366906 Supplier Toronto Research Chemicals Supplier No. TRCD232663100MG
 Begränsad artikel
Beräknad leverans: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 12beta-Deoxycholic Acid Methyl Ester 3alpha-Benzoate
Analyt- eller komponentnamn 12beta-Deoxycholic Acid Methyl Ester 3alpha-Benzoate
Frakt skick Rumstemperatur
Molekylformel C32H46O5
Synonym (3R,5R,8R,9S,10S,12R,13R,14S,17R)-12-Hydroxy-17-((R)-5-methoxy-5-oxopentan-2-yl)-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-3-yl Benzoate
InChI formel InChI=1S/C32H46O5/c1-20(10-15-29(34)36-4)25-13-14-26-24-12-11-22-18-23(37-30(35)21-8-6-5-7-9-21)16-17-31(22,2)27(24)19-28(33)32(25,26)3/h5-9,20,22-28,33H,10-19H2,1-4H3/t20-,22-,23-,24+,25-,26+,27+,28-,31+,32-/m1/s1
Lösningstyp Snyggt
LEDER O=C(O[C@@H]1CC[C@]2(C)[C@@]3([H])C[C@@H](O)[C@]4(C)[C@@H]([C@@H](CCC(OC)=O)C)CC[C@@]4([H])[C@]3([H])CC[C@]2([H])C1)C5=CC=CC=C5
IUPAC-namn (3R,5R,8R,9S,10S,12R,13R,14S,17R)-12-hydroxy-17-((R)-5-metoxoxy-5-oxopentan-2-yl)-10,13-dimetylhexadekahydro-1H-cyklopenta[a]fenantren-3-ylbenzoat
Industry Type Farmaceutisk
Molekylvikt (g/mol) 510.7
Kvantitet 100 mg
Formel vikt 510.33
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.