Läs mer
12Beta-Hydroxyisocholic Acid, TRC
Produktkod. 17961175
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17961175 50 mg
17943405 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17961175 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94348050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 71883-64-2
Molekylformel C24 H40 O5
Molekylvikt (g/mol) 408.57
Synonym (3α,5β,7α,12β)-3,7,12-Trihydroxycholan-24-oic Acid, (R)-4-((3R,5S,7R,8R,9S,10S,12R,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic Acid, 3α,7α,12β-Trihydroxy-5β-cholanic Acid, 3α,7α,12β-Trihydroxy-5β-cholanoic Acid, Lagocholic Acid
IUPAC-namn (4R)-4-[(3R,5S,7R,8R,9S,10S,12R,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@@H](O)[C@]12C
Specifikationer
Kemiskt namn eller material 12Beta-Hydroxyisocholic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20-,22+,23+,24-/m1/s1
Formel vikt 408.2876