Läs mer
13-cis-Retinal 95%, TRC
Produktkod. 30136761
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136761 25 mg
30010059 10 mg
30004297 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136761 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR23990025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 472-86-6
Molekylformel C20 H28 O
Molekylvikt (g/mol) 284.44
IUPAC-namn (2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenal
LEDER C\C(=C\C=O)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C
Specifikationer
Kemiskt namn eller material 13-cis Retinal
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C20H28O/c1-16(8-6-9-17(2)13-15-21)11-12-19-18(3)10-7-14-20(19,4)5/h6,8-9,11-13,15H,7,10,14H2,1-5H3/b9-6+,12-11+,16-8+,17-13-
Formel vikt 284.214
Procent renhet >95%