Läs mer
13-cis-Retinyl Acetate (85%), TRC
Produktkod. 30410497
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30410497 1 mg
1 options
Produktkod. 30410497 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR2399051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 13-cis-Retinyl Acetate
CAS 34356-31-5
Molekylformel C22 H32 O2
Kvantitet 1 mg
Synonym Retinol','15-acetate','13-cis-, Retinol','acetate','13-cis- (8CI,9CI), 13-cis-Retinol acetate, 13-cis-Retinyl acetate, 13-cis-Vitamin A acetate
InChI formel InChI=1S/C22H32O2/c1-17(9-7-10-18(2)14-16-24-20(4)23)12-13-21-19(3)11-8-15-22(21,5)6/h7,9-10,12-14H,8,11,15-16H2,1-6H3/b10-7+,13-12+,17-9+,18-14-
LEDER CC(=O)OC\C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C
IUPAC-namn [(2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
Molekylvikt (g/mol) 328.49
Formel vikt 328.2402